Learn More
Cell Signaling Technology Phospho-Ret (panTyr) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131539
Encompass
The Phospho-Ret (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied free of bovine serum albumin and azide. It is ideal for conjugation with various labels including fluorophores, lanthanides, biotin, and beads.
  • Validated for use in ELISA+ applications.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)).
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of 100 μg each of Ret (C31B4) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody.
  • Human reactivity.
Catalog No. 50-313-1539 Supplier Cell Signaling Technology Supplier No. 84314S
The Phospho-Ret (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes and are supplied free of bovine serum albumin and azide. It is ideal for conjugation with various labels including fluorophores, lanthanides, biotin, and beads.
  • Validated for use in ELISA+ applications.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)).
  • Compatible with platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Consists of 100 μg each of Ret (C31B4) Rabbit Monoclonal Antibody and Phospho-Tyrosine (9E9) Mouse Monoclonal Antibody.
  • Human reactivity.