Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-RIP(S166) Antibody (YA1284) | 10 UL
SDP

Catalog No. 5000441553
Encompass_Preferred

Phospho-RIP(S166) Antibody (YA1284) is a Rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-RIP(S166). It is intended for research use only. This antibody is a crucial tool for investigating TNF-mediated apoptosis, necroptosis, and inflammatory pathways, as RIPK1 (the target protein) plays a central role in these cellular processes.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Mouse
  • Immunogen: A synthetic phospho-peptide corresponding to residues surrounding serine 166 of Mouse RIPK1
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in PBS 59%, sodium azide 0.01%, glycerol 40%, BSA 0.05%
  • Applications: WB, IP, ICC/IF, IHC-P, IHC-F

Catalog No. 50-004-41553 Supplier Medchemexpress LLC Supplier No. HYP8153910UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-RIP(S166) Antibody (YA1284) is a Rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-RIP(S166). It is intended for research use only. This antibody is a crucial tool for investigating TNF-mediated apoptosis, necroptosis, and inflammatory pathways, as RIPK1 (the target protein) plays a central role in these cellular processes.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Mouse
  • Immunogen: A synthetic phospho-peptide corresponding to residues surrounding serine 166 of Mouse RIPK1
  • Sensitivity: Endogenous
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in PBS 59%, sodium azide 0.01%, glycerol 40%, BSA 0.05%
  • Applications: WB, IP, ICC/IF, IHC-P, IHC-F

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.