Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-RIP (Ser320) Antibody, Rabbit, Human, 100 μL
SDP

Catalog No. 502732787
Encompass

The Phospho-RIP (Ser320) Antibody is a rabbit polyclonal antibody that recognizes endogenous levels of RIP protein specifically when phosphorylated at Ser320. This antibody is produced by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding human Ser320 and is subsequently purified via peptide affinity chromatography. It is designed for use in Western blotting applications.

  • Host species: Rabbit
  • Target: RIPK1 (Ser320) phosphate
  • Applications: Western blotting
  • Species reactivity: Human
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol
  • Storage: Store at –20°C

Catalog No. 50-273-2787 Supplier Cell Signaling Technology Supplier No. 58274S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-RIP (Ser320) Antibody is a rabbit polyclonal antibody that recognizes endogenous levels of RIP protein specifically when phosphorylated at Ser320. This antibody is produced by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding human Ser320 and is subsequently purified via peptide affinity chromatography. It is designed for use in Western blotting applications.

  • Host species: Rabbit
  • Target: RIPK1 (Ser320) phosphate
  • Applications: Western blotting
  • Species reactivity: Human
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol
  • Storage: Store at –20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.