Learn More
Cell Signaling Technology Phospho-RIP (Ser415) (F1U7B) Rabbit Monoclonal Antibody, Human, Mouse, 100 μl
SDP
Catalog No. 503133713
Encompass
The Phospho-RIP (Ser415) (F1U7B) Rabbit Monoclonal Antibody is designed to detect endogenous levels of mouse RIP protein specifically when phosphorylated at Ser415. Additionally, this antibody is validated to recognize human RIP protein when phosphorylated at Ser416. It is produced by immunizing animals with a synthetic phosphopeptide corresponding to the residues surrounding Ser415 of the mouse RIP protein.
  • Validated for use in western blotting and immunoprecipitation applications
  • Recognizes endogenous levels of mouse RIP (Ser415) and human RIP (Ser416)
  • Supplied in a stabilized glycerol-based formulation
  • Developed for high specificity as a monoclonal antibody
Catalog No. 50-313-3713 Supplier Cell Signaling Technology Supplier No. 71420S
The Phospho-RIP (Ser415) (F1U7B) Rabbit Monoclonal Antibody is designed to detect endogenous levels of mouse RIP protein specifically when phosphorylated at Ser415. Additionally, this antibody is validated to recognize human RIP protein when phosphorylated at Ser416. It is produced by immunizing animals with a synthetic phosphopeptide corresponding to the residues surrounding Ser415 of the mouse RIP protein.
  • Validated for use in western blotting and immunoprecipitation applications
  • Recognizes endogenous levels of mouse RIP (Ser415) and human RIP (Ser416)
  • Supplied in a stabilized glycerol-based formulation
  • Developed for high specificity as a monoclonal antibody