Learn More
Cell Signaling Technology Phospho-Ron (panTyr) Matched Antibody Pair, Human, ELISA, 1 Kit
SDP
Catalog No. 503131866
Encompass
The Phospho-Ron (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized antibody labeling, such as conjugation with fluorophores, lanthanides, biotin, or beads. It is suitable for platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex. This kit contains capture and detection antibodies that bind to non-overlapping epitopes to support sensitive detection of the target protein.
  • Validated for ELISA and ELISA-like assays.
  • Species reactivity is limited to human.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide.
  • Includes 100 μg each of the capture and detection antibodies.
  • Compatible with diverse labeling chemistries.
Catalog No. 50-313-1866 Supplier Cell Signaling Technology Supplier No. 63895S
The Phospho-Ron (panTyr) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized antibody labeling, such as conjugation with fluorophores, lanthanides, biotin, or beads. It is suitable for platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex. This kit contains capture and detection antibodies that bind to non-overlapping epitopes to support sensitive detection of the target protein.
  • Validated for ELISA and ELISA-like assays.
  • Species reactivity is limited to human.
  • Formulated in 1X PBS (pH 7.8) without BSA or azide.
  • Includes 100 μg each of the capture and detection antibodies.
  • Compatible with diverse labeling chemistries.