Learn More
Cell Signaling Technology Phospho-ROS1 (panTyr) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503131789
Encompass
The Phospho-ROS1 (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating performance in applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and Azide Free formulation, allowing for direct conjugation with labels including fluorophores, lanthanides, biotin, and beads.
  • Validated for ELISA and ELISA-like assays
  • Compatible with diverse labeling chemistries for custom assay development
  • Supplied in 1X PBS (pH 7.8) without BSA or azide additives
  • Suitable for detection of human ROS1 kinase activity and fusion protein biomarkers
Catalog No. 50-313-1789 Supplier Cell Signaling Technology Supplier No. 65355S
The Phospho-ROS1 (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating performance in applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and Azide Free formulation, allowing for direct conjugation with labels including fluorophores, lanthanides, biotin, and beads.
  • Validated for ELISA and ELISA-like assays
  • Compatible with diverse labeling chemistries for custom assay development
  • Supplied in 1X PBS (pH 7.8) without BSA or azide additives
  • Suitable for detection of human ROS1 kinase activity and fusion protein biomarkers