Learn More
ABclonal Technology Phospho-Rpb1 CTD-S2/S5
SDP
Supplier:  ABclonal Technology AP135420UL
Encompass_Preferred
This gene encodes the largest subunit of RNA polymerase II the polymerase responsible for synthesizing messenger RNA in eukaryotes The product of this gene contains a carboxy terminal domain composed of heptapeptide repeats that are essential for polymerase activity These repeats contain serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase In addition this subunit in combination with several other polymerase subunits forms the DNA binding domain of the polymerase a groove in which the DNA template is transcribed into RNA
Catalog No. 50-000-13996