Learn More
Bioss Phospho-RPS6KB1
SDP
Catalog No. 5000065154
Encompass_Preferred
RPS6KB1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65154 Supplier Bioss Supplier No. BSM62970R
RPS6KB1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol