Learn More
Medchemexpress LLC Phospho-RSK1 p90 (Ser380) Antibody (YA158) | 10 UL

Supplier: Medchemexpress LLC HYP8084810UL
Phospho-RSK1 p90 (Ser380) Antibody (YA158) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed for research use. It specifically targets Phospho-RSK1 p90 (Ser380) and is sensitive to endogenous proteins.
- Rabbit host
- Recombinant, monoclonal clonality
- Targets Phospho-RSK1 p90 (Ser380)
- Species reactivity: Human, rat, mouse
- Applications include western blot, immunocytochemistry/immunofluorescence, and immunoprecipitation
- Affinity purified
- Non-conjugated
- Stored at -20°C for 1 year
- Avoid repeated freeze/thaw cycles
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.