Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-RSK1 p90 (Ser380) Antibody (YA158) | 100 UL
SDP

Catalog No. 5000441955
Encompass_Preferred

Phospho-RSK1 p90 (Ser380) Antibody (YA158) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Phospho-RSK1 p90 (Ser380). It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Observed molecular weight of 90 kDa
  • Human, rat, mouse species reactivity
  • Affinity purified
  • Non-conjugated
  • Supplied in liquid form
  • Stored at -20°C for 1 year

Catalog No. 50-004-41955 Supplier Medchemexpress LLC Supplier No. HYP80848100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-RSK1 p90 (Ser380) Antibody (YA158) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Phospho-RSK1 p90 (Ser380). It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Observed molecular weight of 90 kDa
  • Human, rat, mouse species reactivity
  • Affinity purified
  • Non-conjugated
  • Supplied in liquid form
  • Stored at -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.