Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-RSK1 p90 (Ser380) Antibody (YA158) | 50 UL
SDP

Catalog No. 5000442208
Encompass_Preferred

Phospho-RSK1 p90 (Ser380) Antibody (YA158) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-RSK1 p90 (Ser380). It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Reacts with human, rat, and mouse species
  • Immunogen is a synthetic phosphopeptide
  • Non-conjugated antibody
  • Phosphorylated modification
  • IgG isotype
  • Supplied in phosphate buffered saline with BSA, sodium azide, and glycerol
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Immunoprecipitation

Catalog No. 50-004-42208 Supplier Medchemexpress LLC Supplier No. HYP8084850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-RSK1 p90 (Ser380) Antibody (YA158) is a rabbit-derived, non-conjugated IgG monoclonal antibody specifically targeting Phospho-RSK1 p90 (Ser380). It is intended for research use only.

  • Rabbit host
  • Recombinant, monoclonal clonality
  • Reacts with human, rat, and mouse species
  • Immunogen is a synthetic phosphopeptide
  • Non-conjugated antibody
  • Phosphorylated modification
  • IgG isotype
  • Supplied in phosphate buffered saline with BSA, sodium azide, and glycerol
  • Store at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Immunoprecipitation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.