Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-RSK1 p90 (Thr359/Ser363) antibody (YA157) | 100 UL
SDP

Catalog No. 5000442209
Encompass_Preferred

This Phospho-RSK1 p90 (Thr359/Ser363) antibody (YA157) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It is designed for research applications such as Western Blot (WB) and Immunoprecipitation (IP).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 0.3 mg/mL
  • Application: WB, IP
  • Dilution ratio: WB: 1:500-1:1000; IP: 1:20
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% sodium azide as preservative
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Predicted molecular weight: 83 kDa
  • Observed molecular weight: 90 kDa

Catalog No. 50-004-42209 Supplier Medchemexpress LLC Supplier No. HYP80849100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Phospho-RSK1 p90 (Thr359/Ser363) antibody (YA157) is a rabbit-derived, non-conjugated IgG monoclonal antibody. It is designed for research applications such as Western Blot (WB) and Immunoprecipitation (IP).

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Concentration: 0.3 mg/mL
  • Application: WB, IP
  • Dilution ratio: WB: 1:500-1:1000; IP: 1:20
  • Formulation: Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% sodium azide as preservative
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Predicted molecular weight: 83 kDa
  • Observed molecular weight: 90 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.