Learn More
Abcam Phospho S232 PDH E1 alpha protein (PDHA1) Profiling ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1153431X96TESTS
Encompass_Preferred
Phospho S232 PDH E1 alpha protein (PDHA1) Profiling ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Phospho S232 PDH E1 alpha protein (PDHA1) Profiling in Cell culture extracts, Tissue Extracts samples from a wide range of species.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1325273