Learn More
Cell Signaling Technology Phospho-S6 Ribosomal Protein (Ser235/236) (E2R1O) Mouse mAb | 100ul
SDP
Supplier:  Cell Signaling Technology 62016S
Encompass
Phospho-S6 Ribosomal Protein (Ser235/236) (E2R1O) Mouse mAb detects endogenous levels of S6 ribosomal protein only when phosphorylated at Ser235 and Ser236.
Catalog No. NC1592057