Learn More
ECM Bioscience Anti-Phosphoserine/threonine Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM3801
Encompass_Preferred
PM3801 Phosphoserine/threonine is a mix of two clones: Clone M380A was generated from a phosphothreonine synthetic peptide (coupled to carrier protein) and Clone M380B was generated.from a phosphoserine synthetic peptide (coupled to carrier protein). Accession #. The antibody detects many serine or threonine phosphorylated proteins by western blot, immunocytochemistry, and ELISA. Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = phosphotyrosine, phoshoserine, phoshothreonine
Catalog No. 50-172-0851