Learn More
Abcam Phospho-SHP-1 (S591) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2799231X96TESTS
Encompass_Preferred
Phospho-SHP-1 (S591) ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-SHP-1 (S591) in Human in Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1956912