Learn More
BIOHIPPO Phospho-SIRT1 Ser47 rAb W-R71
SDP
Catalog No. 5000975514
Encompass
U-Blot Phospho-SIRT1(Ser47) Rabbit mAb is a Rabbit-derived recombinant monoclonal antibody (clone W-R71) that specifically recognizes Phospho-SIRT1 (Ser47) a phosphorylation modification at Ser47 It shows reactivity with Human Validated applications include WB IHC-P Recommended dilutions WB 1500-11000 IHC-P 150-1200 Purified by protein a SIRT1 is a member of the Silent Information Regulator2 (SIR2) family which is a highly conserved group of genes that encode nicotinamide adenine dinucleotide (NAD)-dependent protein deacetylases also known as class III histone deacetylases The first discovered and best characterized of these genes is Saccharomyces cerevisiae SIR2 which is involved in silencing of mating type loci telomere maintenance DNA damage response and cell aging Supplied as 100 uL
Catalog No. 50-009-75514 Supplier BIOHIPPO Supplier No. BHA21821385
U-Blot Phospho-SIRT1(Ser47) Rabbit mAb is a Rabbit-derived recombinant monoclonal antibody (clone W-R71) that specifically recognizes Phospho-SIRT1 (Ser47) a phosphorylation modification at Ser47 It shows reactivity with Human Validated applications include WB IHC-P Recommended dilutions WB 1500-11000 IHC-P 150-1200 Purified by protein a SIRT1 is a member of the Silent Information Regulator2 (SIR2) family which is a highly conserved group of genes that encode nicotinamide adenine dinucleotide (NAD)-dependent protein deacetylases also known as class III histone deacetylases The first discovered and best characterized of these genes is Saccharomyces cerevisiae SIR2 which is involved in silencing of mating type loci telomere maintenance DNA damage response and cell aging Supplied as 100 uL