Learn More
Cell Signaling Technology Phospho-SMAD2 Monoclonal Antibody, Rabbit, Human, 100 μg
SDP
Catalog No. 503133309
Encompass
This rabbit monoclonal antibody specifically detects endogenous levels of SMAD2 only when dually phosphorylated at serines 465 and 467. The antibody is produced by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Ser465/467 of human SMAD2. This carrier-free formulation is ideal for custom antibody labeling technologies, including conjugation to fluorophores, metals, lanthanides, and oligonucleotides.
  • Specifically detects SMAD2 phosphorylated at Ser465 and 467
  • BSA and azide free for versatile downstream labeling applications
  • Quality control tested by size exclusion chromatography to ensure antibody integrity
  • Validated for use in ELISA-like assays
  • Recombinant rabbit monoclonal antibody
Catalog No. 50-313-3309 Supplier Cell Signaling Technology Supplier No. 81000SF
This rabbit monoclonal antibody specifically detects endogenous levels of SMAD2 only when dually phosphorylated at serines 465 and 467. The antibody is produced by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Ser465/467 of human SMAD2. This carrier-free formulation is ideal for custom antibody labeling technologies, including conjugation to fluorophores, metals, lanthanides, and oligonucleotides.
  • Specifically detects SMAD2 phosphorylated at Ser465 and 467
  • BSA and azide free for versatile downstream labeling applications
  • Quality control tested by size exclusion chromatography to ensure antibody integrity
  • Validated for use in ELISA-like assays
  • Recombinant rabbit monoclonal antibody