Learn More
Cell Signaling Technology Phospho-SMAD2 (Ser465/467) Matched Antibody Pair, Human, Mouse, Monkey, 1 Kit
SDP
Catalog No. 503134154
Encompass
The Phospho-SMAD2 (Ser465/467) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, such as fluorophores, lanthanides, biotin, or beads. This antibody pair includes capture and detection antibodies that bind to non-overlapping epitopes. It is compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Supplied as a kit containing 100 μg each of mouse monoclonal antibody and rabbit monoclonal antibody
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, 140 mM NaCl)
  • BSA and azide free for easy labeling
  • Validated for reactivity in human, mouse, and monkey species
  • Ideal for development of high-throughput ELISA assays
Catalog No. 50-313-4154 Supplier Cell Signaling Technology Supplier No. 71460S
The Phospho-SMAD2 (Ser465/467) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling, such as fluorophores, lanthanides, biotin, or beads. This antibody pair includes capture and detection antibodies that bind to non-overlapping epitopes. It is compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Supplied as a kit containing 100 μg each of mouse monoclonal antibody and rabbit monoclonal antibody
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, 140 mM NaCl)
  • BSA and azide free for easy labeling
  • Validated for reactivity in human, mouse, and monkey species
  • Ideal for development of high-throughput ELISA assays