Learn More
Cell Signaling Technology Phospho-SMAD3 Matched Antibody Pair, Human, Mouse, Mink, 1 Kit
SDP
Catalog No. 503132131
Encompass
The Phospho-SMAD3 (Ser423/425) Matched Antibody Pair is designed for high-throughput immunoassay platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating sensitive detection of SMAD3 phosphorylated at Ser423/425. It is compatible with a range of conjugation technologies, including fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF)
  • Includes capture and detection antibodies validated for immunoassay performance
  • Supplied in a BSA and azide-free formulation
  • Validated for reactivity in human, mouse, and mink species
  • Suitable for research use only
Catalog No. 50-313-2131 Supplier Cell Signaling Technology Supplier No. 10496S
The Phospho-SMAD3 (Ser423/425) Matched Antibody Pair is designed for high-throughput immunoassay platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, facilitating sensitive detection of SMAD3 phosphorylated at Ser423/425. It is compatible with a range of conjugation technologies, including fluorophores, lanthanides, biotin, and beads.
  • Compatible with platforms such as MSD, Quanterix Simoa, Luminex, and Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF)
  • Includes capture and detection antibodies validated for immunoassay performance
  • Supplied in a BSA and azide-free formulation
  • Validated for reactivity in human, mouse, and mink species
  • Suitable for research use only