Learn More
Meso Scale Discovery Phospho-STAT Panel Kit
SDP
Catalog No. 5000078777
Encompass
The Meso Scale Discovery Phospho-STAT Panel Kit is a multiplex assay that measures the levels of phosphorylated STAT1 STAT3 STAT5 and STAT6 in cell lysates The assay uses MSD s electrochemiluminescent detection technology and provides a rapid and sensitive way to monitor STAT protein activation and signaling The kit includes a 96-well plate coated with capture antibodies specific for the phosphorylated STAT proteins It also contains detection antibodies and reagents needed to perform the assay Results can be read on MSD s SECTOR Imager instrument The kit allows for the simultaneous measurement of multiple STAT proteins from a single sample
Catalog No. 50-000-78777 Supplier Meso Scale Discovery Supplier No. K15202D1
The Meso Scale Discovery Phospho-STAT Panel Kit is a multiplex assay that measures the levels of phosphorylated STAT1 STAT3 STAT5 and STAT6 in cell lysates The assay uses MSD s electrochemiluminescent detection technology and provides a rapid and sensitive way to monitor STAT protein activation and signaling The kit includes a 96-well plate coated with capture antibodies specific for the phosphorylated STAT proteins It also contains detection antibodies and reagents needed to perform the assay Results can be read on MSD s SECTOR Imager instrument The kit allows for the simultaneous measurement of multiple STAT proteins from a single sample