Learn More
Meso Scale Discovery Phospho-STAT Panel Kit
SDP
Catalog No. 5000078925
Encompass
The Meso Scale Discovery Phospho-STAT Panel Kit is a multiplex immunoassay that simultaneously measures the levels of phosphorylated STAT1 STAT3 STAT5 and STAT6 in cell lysates The assay uses MSD s electrochemiluminescent detection technology and provides a rapid and sensitive way to monitor STAT protein activation and signaling through cytokine and growth factor receptors The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrator It can process up to 80 samples in duplicate and has a total assay time of 3 5 hours The kit is for research use only
Catalog No. 50-000-78925 Supplier Meso Scale Discovery Supplier No. K15202D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho-STAT Panel Kit is a multiplex immunoassay that simultaneously measures the levels of phosphorylated STAT1 STAT3 STAT5 and STAT6 in cell lysates The assay uses MSD s electrochemiluminescent detection technology and provides a rapid and sensitive way to monitor STAT protein activation and signaling through cytokine and growth factor receptors The kit includes a 96-well plate coated with capture antibodies detection antibodies and calibrator It can process up to 80 samples in duplicate and has a total assay time of 3 5 hours The kit is for research use only