Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-Stat1 (Ser727) Rabbit Monoclonal Antibody, Recombinant, Unconjugated, 20 μL
SDP

Catalog No. 502730410
Encompass

This phospho-specific rabbit monoclonal antibody recognizes endogenous levels of Stat1 protein only when phosphorylated at Ser727. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding the Ser727 site of the human Stat1 protein. It is validated for multiple applications, including Western blotting, immunoprecipitation, immunohistochemistry, chromatin immunoprecipitation, and CUT&RUN assays.

  • Host species: Rabbit
  • Target modification: Phosphate at Ser727
  • Species reactivity: Human, mouse, rat, and monkey
  • Validated applications: Western blotting, immunoprecipitation, immunohistochemistry (paraffin), chromatin IP, and CUT&RUN
  • Supplied in: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage conditions: Store at –20°C; do not aliquot

Catalog No. 50-273-0410 Supplier Cell Signaling Technology Supplier No. 8826T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This phospho-specific rabbit monoclonal antibody recognizes endogenous levels of Stat1 protein only when phosphorylated at Ser727. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding the Ser727 site of the human Stat1 protein. It is validated for multiple applications, including Western blotting, immunoprecipitation, immunohistochemistry, chromatin immunoprecipitation, and CUT&RUN assays.

  • Host species: Rabbit
  • Target modification: Phosphate at Ser727
  • Species reactivity: Human, mouse, rat, and monkey
  • Validated applications: Western blotting, immunoprecipitation, immunohistochemistry (paraffin), chromatin IP, and CUT&RUN
  • Supplied in: 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage conditions: Store at –20°C; do not aliquot

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.