Learn More
Meso Scale Discovery Phospho-STAT3 (Tyr705) Kit, 96-well plate, 25 plates, detects STAT3 pTyr705, suitable for human, mouse, rat samples, compatible with various MSD instruments
SDP
Catalog No. NC3072029
Encompass
The MESO SCALE DISCOVERY Phospho-STAT3 (Tyr705) Kit is designed as a rapid and convenient assay for the detection of STAT3 phosphorylated at tyrosine 705. The kit is suitable for research applications focused on phosphoproteins and intracellular signaling across various species, including human, mouse, and rat. Utilizing a 96-well plate format, it enables efficient processing of samples with high-throughput compatibility.
  • Detects STAT3 phosphorylated at Tyr705, a crucial activation site.
  • Validated for use with human, mouse, and rat samples.
  • Compatible with multiple MSD instruments.
  • Contains all essential reagents for accurate results.
  • Includes comprehensive kit contents such as SULFO-TAG antibody and various buffers.
  • Intended for research use only.
  • Stored as multi-component modules.
Catalog No. NC3072029 Supplier Meso Scale Discovery Supplier No. K150SVD4
Add to Cart
Add to Cart
Request a Quote
The MESO SCALE DISCOVERY Phospho-STAT3 (Tyr705) Kit is designed as a rapid and convenient assay for the detection of STAT3 phosphorylated at tyrosine 705. The kit is suitable for research applications focused on phosphoproteins and intracellular signaling across various species, including human, mouse, and rat. Utilizing a 96-well plate format, it enables efficient processing of samples with high-throughput compatibility.
  • Detects STAT3 phosphorylated at Tyr705, a crucial activation site.
  • Validated for use with human, mouse, and rat samples.
  • Compatible with multiple MSD instruments.
  • Contains all essential reagents for accurate results.
  • Includes comprehensive kit contents such as SULFO-TAG antibody and various buffers.
  • Intended for research use only.
  • Stored as multi-component modules.