Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-Stat5a (Tyr694) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP

Catalog No. 503134325
Encompass

The Phospho-Stat5a (Tyr694) Matched Antibody Pair is designed for use in immunoassays and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for precise detection of phosphorylated Stat5a (Tyr694) without cross-reactivity with phospho-Stat5b (Tyr699).

  • Compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as BSA and azide free for optimal performance in downstream labeling.
  • Validated for human species reactivity.
  • Contains 100 μg each of capture and detection monoclonal antibodies.
  • Storage at -20°C recommended in single-use aliquots.

Catalog No. 50-313-4325 Supplier Cell Signaling Technology Supplier No. 79167S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-Stat5a (Tyr694) Matched Antibody Pair is designed for use in immunoassays and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, allowing for precise detection of phosphorylated Stat5a (Tyr694) without cross-reactivity with phospho-Stat5b (Tyr699).

  • Compatible with various conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Suitable for platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as BSA and azide free for optimal performance in downstream labeling.
  • Validated for human species reactivity.
  • Contains 100 μg each of capture and detection monoclonal antibodies.
  • Storage at -20°C recommended in single-use aliquots.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.