Learn More
Cell Signaling Technology Phospho-Stat6 (Tyr641) Matched Antibody Pair, Unconjugated, Human, 1 Kit
SDP
Catalog No. 503132353
Encompass
The Phospho-Stat6 (Tyr641) Matched Antibody Pair II is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, enabling versatility for conjugation with fluorophores, lanthanides, biotin, or beads.
  • Ideal for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)) and are BSA and azide free.
  • Consists of 100 μg each of rabbit monoclonal capture and detection antibodies.
  • Optimized for human species reactivity.
  • Validated for ELISA and ELISA-like assays.
  • Recommended for storage at -20°C in single-use aliquots.
Catalog No. 50-313-2353 Supplier Cell Signaling Technology Supplier No. 93674S
The Phospho-Stat6 (Tyr641) Matched Antibody Pair II is designed for immunoassay technologies and high-throughput ELISA platforms that require specialized or custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, enabling versatility for conjugation with fluorophores, lanthanides, biotin, or beads.
  • Ideal for platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)) and are BSA and azide free.
  • Consists of 100 μg each of rabbit monoclonal capture and detection antibodies.
  • Optimized for human species reactivity.
  • Validated for ELISA and ELISA-like assays.
  • Recommended for storage at -20°C in single-use aliquots.