Learn More
Bioss Phospho-Syk- Tyr323 Recombinan
SDP
Catalog No. 5000066189
Encompass_Preferred
Syk Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66189 Supplier Bioss Supplier No. BSM62848R
Syk Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol