Learn More
Cell Signaling Technology Matched Antibody Pair, Phospho-Tau, Human, Mouse, Rat, 1 Kit
SDP
Catalog No. 503134122
Encompass
The Phospho-Tau (Thr205) Matched Antibody Pair provides essential components for the sensitive detection of tau phosphorylated at Thr205 in diverse immunoassay and high-throughput platforms. This kit includes capture and detection rabbit monoclonal antibodies that bind to non-overlapping epitopes, enabling robust sandwich-based detection of endogenous protein expression. It is suitable for applications such as ELISA, and is compatible with specialized labeling technologies including lanthanides, fluorophores, biotin, and beads.
  • Includes high-affinity capture and detection rabbit monoclonal antibodies.
  • Supplied in BSA and azide-free formulation to support custom conjugation.
  • Validated for reactivity with human, mouse, and rat samples.
  • Optimized for sandwich immunoassay technologies including ELISA, MSD, Quanterix Simoa, and Alpha Technology platforms.
  • Stable liquid formulation.
Catalog No. 50-313-4122 Supplier Cell Signaling Technology Supplier No. 66673S
The Phospho-Tau (Thr205) Matched Antibody Pair provides essential components for the sensitive detection of tau phosphorylated at Thr205 in diverse immunoassay and high-throughput platforms. This kit includes capture and detection rabbit monoclonal antibodies that bind to non-overlapping epitopes, enabling robust sandwich-based detection of endogenous protein expression. It is suitable for applications such as ELISA, and is compatible with specialized labeling technologies including lanthanides, fluorophores, biotin, and beads.
  • Includes high-affinity capture and detection rabbit monoclonal antibodies.
  • Supplied in BSA and azide-free formulation to support custom conjugation.
  • Validated for reactivity with human, mouse, and rat samples.
  • Optimized for sandwich immunoassay technologies including ELISA, MSD, Quanterix Simoa, and Alpha Technology platforms.
  • Stable liquid formulation.