Learn More
Cell Signaling Technology Phospho-TFEB (Ser122) Matched Antibody Pair, Human, 1 Kit
SDP
Catalog No. 503132337
Encompass
The Phospho-TFEB (Ser122) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications utilizing fluorophores, lanthanides, biotin, or beads.
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (e.g., AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Validated for human species reactivity.
  • Supplied in a BSA and azide-free formulation in 1X PBS (pH 7.8).
  • Kit components include 100 μg of Phospho-TFEB (Ser122) rabbit monoclonal antibody and 100 μg of TFEB rabbit monoclonal antibody.
  • Recommended for research use only.
Catalog No. 50-313-2337 Supplier Cell Signaling Technology Supplier No. 26770S
The Phospho-TFEB (Ser122) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom antibody labeling. This kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications utilizing fluorophores, lanthanides, biotin, or beads.
  • Compatible with diverse platforms including MSD, Quanterix Simoa, Luminex, and Alpha Technology (e.g., AlphaScreen, AlphaLISA, LANCE, HTRF).
  • Validated for human species reactivity.
  • Supplied in a BSA and azide-free formulation in 1X PBS (pH 7.8).
  • Kit components include 100 μg of Phospho-TFEB (Ser122) rabbit monoclonal antibody and 100 μg of TFEB rabbit monoclonal antibody.
  • Recommended for research use only.