Learn More
Enzo Life Sciences Phosphothreonine detection set (1Set)
SDP
Supplier:  Enzo Life Sciences ALX850024KI01
Encompass_Preferred
This kit offers a set of three different monoclonal antibodies specific for threonine phosphorylation sites. Phosphorylation patterns in a given cell extract may differ when probed with different antibodies due to sequence specificity. Recognition therefore depends on two criteria: Phosphorylation and surrounding aa motif. If one of the two criteria is not fulfilled, the antibody will not detect the phosphorylation site. Long Term Storage: -20°C. Assay Time: Overnight + 2 hours. Applications: IP, WB .
Catalog No. 50-201-0096