Learn More
Meso Scale Discovery Phospho/Total MEK1/2 WCL5
SDP
Catalog No. 5000080287
Encompass
The Meso Scale Discovery Phospho/Total MEK1/2 Whole Cell Lysate Kit is designed to monitor MEK1/2 phosphorylation and expression in cell signaling studies The kit includes a 96-well plate coated with capture antibodies against total MEK1/2 and phospho-MEK1/2 at Ser217/221 The kit provides all necessary reagents for cell lysis and detection using an electrochemiluminescent readout This allows for high sensitivity quantitation of phospho and total levels of MEK1/2 from cell lysates The kit can be used to study MEK1/2 activation in response to growth factors cytokines and pharmacological MEK inhibitors
Catalog No. 50-000-80287 Supplier Meso Scale Discovery Supplier No. K15129D2
The Meso Scale Discovery Phospho/Total MEK1/2 Whole Cell Lysate Kit is designed to monitor MEK1/2 phosphorylation and expression in cell signaling studies The kit includes a 96-well plate coated with capture antibodies against total MEK1/2 and phospho-MEK1/2 at Ser217/221 The kit provides all necessary reagents for cell lysis and detection using an electrochemiluminescent readout This allows for high sensitivity quantitation of phospho and total levels of MEK1/2 from cell lysates The kit can be used to study MEK1/2 activation in response to growth factors cytokines and pharmacological MEK inhibitors