Learn More
Meso Scale Discovery Phospho/Total p38 WCL20
SDP
Catalog No. 5000078894
Encompass
The Meso Scale Discovery Phospho (Thr180/Tyr182)/Total p38 Whole Cell Lysate Kit (K15112D) is designed to measure endogenous levels of phospho and total p38 in whole cell lysates The kit includes a 96-well plate coated with capture antibodies against phospho and total p38 detection antibodies against phospho and total p38 read buffer wash buffer blocking solution cell lysis buffer and a plate seal The assay can detect phospho and total p38 with a typical sensitivity of 50-100 g/mL lysate This kit allows for the quantitative measurement of p38 phosphorylation and expression from cell lysates
Catalog No. 50-000-78894 Supplier Meso Scale Discovery Supplier No. K15112D3
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho (Thr180/Tyr182)/Total p38 Whole Cell Lysate Kit (K15112D) is designed to measure endogenous levels of phospho and total p38 in whole cell lysates The kit includes a 96-well plate coated with capture antibodies against phospho and total p38 detection antibodies against phospho and total p38 read buffer wash buffer blocking solution cell lysis buffer and a plate seal The assay can detect phospho and total p38 with a typical sensitivity of 50-100 g/mL lysate This kit allows for the quantitative measurement of p38 phosphorylation and expression from cell lysates