Learn More
Bioss Phospho-TRIM28 - Ser824 Monocl
SDP
Catalog No. 5000065103
Encompass_Preferred
TRIM28 (Ser824) Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65103 Supplier Bioss Supplier No. BSM63001R
TRIM28 (Ser824) Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol