Learn More
Enzo Life Sciences Phosphotyrosine monoclonal antibody (16F4) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804174C100
Encompass_Preferred
Clone: 16F4. Isotype: IgG1. Immunogen: Phosphotyrosine containing peptides conjugated to KLH. Species reactivity: Species independent. Specificity: Epitope:.DIpYAE.Recognizes phosphotyrosine in the context of the surrounding amino acids, tolerating hydrophobic amino acids directly neighbouring the phosphotyrosine. Applications: ELISA, IP, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9815