Learn More
Cell Signaling Technology Matched Antibody Pair, Phospho-Tyro3 (panTyr), Human, Unconjugated, 1 Kit
SDP
Catalog No. 503133386
Encompass
The Phospho-Tyro3 (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring specialized antibody labeling. The kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation.
  • Ideal for customized antibody labeling with fluorophores, lanthanides, biotin, or beads
  • Optimized for high-throughput ELISA and similar immunoassay platforms
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • BSA and azide free for maximum versatility in downstream applications
  • Includes capture and detection antibodies
Catalog No. 50-313-3386 Supplier Cell Signaling Technology Supplier No. 51735S
The Phospho-Tyro3 (panTyr) Matched Antibody Pair is designed for immunoassay technologies and high-throughput ELISA platforms requiring specialized antibody labeling. The kit includes capture and detection antibodies that bind to non-overlapping epitopes, making it suitable for applications such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. The antibodies are provided in a BSA and azide-free formulation.
  • Ideal for customized antibody labeling with fluorophores, lanthanides, biotin, or beads
  • Optimized for high-throughput ELISA and similar immunoassay platforms
  • Supplied in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl, pH 7.8)
  • BSA and azide free for maximum versatility in downstream applications
  • Includes capture and detection antibodies