Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Tyrosine Antibody(YA3552) | 100 UL
SDP

Catalog No. 5000444161
Encompass_Preferred

Phospho-Tyrosine Antibody(YA3552) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Phospho-Tyrosine. Tyrosine phosphorylation plays a key role in cellular signaling. Research studies have shown that in cancer, unregulated tyrosine kinase activity can drive malignancy and tumor formation by generating inappropriate proliferation and survival signals. Antibodies specific for phospho-tyrosine have been invaluable reagents in these studies.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets phospho-tyrosine
  • Recombinant
  • Reacts with human, mouse, and rat species
  • Immunogen is a synthesized peptide
  • IgG isotype
  • Liquid appearance
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • For research use only

Catalog No. 50-004-44161 Supplier Medchemexpress LLC Supplier No. HYP83749100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-Tyrosine Antibody(YA3552) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to Phospho-Tyrosine. Tyrosine phosphorylation plays a key role in cellular signaling. Research studies have shown that in cancer, unregulated tyrosine kinase activity can drive malignancy and tumor formation by generating inappropriate proliferation and survival signals. Antibodies specific for phospho-tyrosine have been invaluable reagents in these studies.

  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets phospho-tyrosine
  • Recombinant
  • Reacts with human, mouse, and rat species
  • Immunogen is a synthesized peptide
  • IgG isotype
  • Liquid appearance
  • Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Shipped with blue ice
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.