Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Tyrosine Antibody(YA3552) | 50 UL
SDP

Catalog No. 5000443431
Encompass_Preferred

Phospho-Tyrosine Antibody(YA3552) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-Tyrosine. This antibody is suitable for research use only.

  • Host: Rabbit
  • Clonality: Monoclonal, recombinant
  • Species reactivity: Human, mouse, rat
  • Immunogen: A synthesized peptide derived from Phosphotyrosine XXXX-pY-XXXX
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200), IP (1:50), FC (1:50-1:100)

Catalog No. 50-004-43431 Supplier Medchemexpress LLC Supplier No. HYP8374950UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phospho-Tyrosine Antibody(YA3552) is a Rabbit-derived, non-conjugated IgG monoclonal antibody that targets Phospho-Tyrosine. This antibody is suitable for research use only.

  • Host: Rabbit
  • Clonality: Monoclonal, recombinant
  • Species reactivity: Human, mouse, rat
  • Immunogen: A synthesized peptide derived from Phosphotyrosine XXXX-pY-XXXX
  • Conjugation: Non-conjugated
  • Modification: Phosphorylated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 10mM PBS, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Shipping: Shipped with blue ice
  • Applications: WB (1:500-1:1000), ICC/IF (1:50-1:200), IP (1:50), FC (1:50-1:100)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.