Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-Ubiquitin (Ser57) Rabbit Monoclonal Antibody, Human, Western Blotting, 100 μL
SDP

Catalog No. 503134084
Encompass

The Phospho-Ubiquitin (Ser57) (F1I4R) Rabbit Monoclonal Antibody is a recombinant antibody designed to detect ubiquitin when phosphorylated at Ser57. It is generated by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Ser57 of the human ubiquitin protein. This antibody is validated for western blotting and specifically recognizes transfected levels of the target protein.

  • Host species: Rabbit
  • Reactivity: Human
  • Application: Validated for western blotting at 1:1,000 dilution
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage: Store at -20°C; do not aliquot the antibody
  • Performance: Superior lot-to-lot consistency and continuous supply

Catalog No. 50-313-4084 Supplier Cell Signaling Technology Supplier No. 88056S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Phospho-Ubiquitin (Ser57) (F1I4R) Rabbit Monoclonal Antibody is a recombinant antibody designed to detect ubiquitin when phosphorylated at Ser57. It is generated by immunizing animals with a synthetic phosphopeptide corresponding to residues surrounding Ser57 of the human ubiquitin protein. This antibody is validated for western blotting and specifically recognizes transfected levels of the target protein.

  • Host species: Rabbit
  • Reactivity: Human
  • Application: Validated for western blotting at 1:1,000 dilution
  • Formulation: Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide
  • Storage: Store at -20°C; do not aliquot the antibody
  • Performance: Superior lot-to-lot consistency and continuous supply

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.