Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-Ubiquitin (Ser65) (E5T1W) Rabbit Monoclonal Antibody, Human, 100 μL
SDP

Catalog No. 502731880
Encompass

This recombinant rabbit monoclonal antibody is designed to specifically detect endogenous levels of human ubiquitin protein only when it is phosphorylated at Ser65. The antibody is produced by immunizing animals with a synthetic phospho-peptide corresponding to residues surrounding Ser65 of human ubiquitin, a conserved polypeptide essential for the ubiquitin-proteasome pathway.

  • Specifically recognizes phosphorylated ubiquitin (Ser65)
  • Validated for immunofluorescence and flow cytometry applications
  • Recombinant rabbit monoclonal antibody
  • Species reactivity confirmed in human
  • Predicted to react with mouse and rat models based on 100% sequence homology

Catalog No. 50-273-1880 Supplier Cell Signaling Technology Supplier No. 70973S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This recombinant rabbit monoclonal antibody is designed to specifically detect endogenous levels of human ubiquitin protein only when it is phosphorylated at Ser65. The antibody is produced by immunizing animals with a synthetic phospho-peptide corresponding to residues surrounding Ser65 of human ubiquitin, a conserved polypeptide essential for the ubiquitin-proteasome pathway.

  • Specifically recognizes phosphorylated ubiquitin (Ser65)
  • Validated for immunofluorescence and flow cytometry applications
  • Recombinant rabbit monoclonal antibody
  • Species reactivity confirmed in human
  • Predicted to react with mouse and rat models based on 100% sequence homology

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.