Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-ULK1 Monoclonal Antibody, Rabbit, 100 μL
SDP

Catalog No. 502730323
Encompass

This antibody is a recombinant rabbit monoclonal antibody developed to detect endogenous levels of ULK1 protein specifically when phosphorylated at Ser317. It is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding the Ser317 site of human ULK1. This reagent is suitable for applications including Western blotting and immunoprecipitation to study autophagy signaling and protein kinase regulation.

  • Validated for Western blotting and immunoprecipitation applications.
  • Specifically recognizes ULK1 phosphorylated at Ser317.
  • Host species is rabbit.
  • Provided in a stable formulation with 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.
  • Recombinant antibody production ensures consistency.
  • Species reactivity confirmed for human, mouse, and rat.

Catalog No. 50-273-0323 Supplier Cell Signaling Technology Supplier No. 89267S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody is a recombinant rabbit monoclonal antibody developed to detect endogenous levels of ULK1 protein specifically when phosphorylated at Ser317. It is generated by immunizing animals with a synthetic peptide corresponding to residues surrounding the Ser317 site of human ULK1. This reagent is suitable for applications including Western blotting and immunoprecipitation to study autophagy signaling and protein kinase regulation.

  • Validated for Western blotting and immunoprecipitation applications.
  • Specifically recognizes ULK1 phosphorylated at Ser317.
  • Host species is rabbit.
  • Provided in a stable formulation with 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, and 50% glycerol.
  • Recombinant antibody production ensures consistency.
  • Species reactivity confirmed for human, mouse, and rat.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.