Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phospho-Vimentin (Ser56) Antibody (YA138) | 100 UL
SDP

Supplier:  Medchemexpress LLC HYP80863100UL

Encompass_Preferred

Phospho-Vimentin (Ser56) antibody (YA138) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, specifically targeting Phospho-Vimentin (Ser56) for research applications. Vimentins, as class-III intermediate filaments, are present in various non-epithelial cells, particularly mesenchymal cells. They are integral to cellular structure, attaching to the nucleus, endoplasmic reticulum, and mitochondria. This antibody can help in understanding vimentin's role in cell directional movement, orientation, cell sheet organization, and Golgi complex polarization during cell migration.

  • Host: Rabbit
  • Clonality: Recombinant, monoclonal
  • Species reactivity: Human
  • Applications: WB, ICC/IF, IP
  • Subcellular localization: Cytoplasm, cytoskeleton, nucleus matrix, cell membrane
  • Storage and stability: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles

Catalog No. 50-004-42212


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.