Learn More
Medchemexpress LLC Phospho-Vimentin (Ser56) Antibody (YA138) | 50 UL
SDP
Catalog No. 5000442506
Encompass_Preferred
Phospho-Vimentin (Ser56) Antibody (YA138) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Vimentin (Ser56). It is supplied as a liquid and is affinity purified. Vimentin is a type III intermediate filament protein involved in maintaining cell shape, cytoplasm integrity, and stabilizing cytoskeletal interactions.
  • Rabbit-derived IgG monoclonal antibody
  • Targets Phospho-Vimentin (Ser56)
  • Used in Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP) applications
  • Reactivity with Human species
  • Stable for 1 year when stored at -20°C
Catalog No. 50-004-42506 Supplier Medchemexpress LLC Supplier No. HYP8086350UL
Phospho-Vimentin (Ser56) Antibody (YA138) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting Phospho-Vimentin (Ser56). It is supplied as a liquid and is affinity purified. Vimentin is a type III intermediate filament protein involved in maintaining cell shape, cytoplasm integrity, and stabilizing cytoskeletal interactions.
  • Rabbit-derived IgG monoclonal antibody
  • Targets Phospho-Vimentin (Ser56)
  • Used in Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Immunoprecipitation (IP) applications
  • Reactivity with Human species
  • Stable for 1 year when stored at -20°C