Learn More
Cell Signaling Technology Phospho-YAP (Ser397) Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503133263
Encompass
The Phospho-YAP (Ser397) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom or specialized antibody labeling, such as fluorophores, lanthanides, beads, or biotin. This kit contains capture and detection antibodies that bind to non-overlapping epitopes and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of Phospho-YAP (Ser397) (D1E7Y) rabbit monoclonal antibody and YAP (1A12) mouse monoclonal antibody.
  • Formulated in 1X PBS (pH 7.8), free of BSA and azide.
  • Optimized for human species reactivity.
  • Supplied as a liquid in a clear appearance.
  • Recommended for use in ELISA and related immunoassay applications.
Catalog No. 50-313-3263 Supplier Cell Signaling Technology Supplier No. 59695S
The Phospho-YAP (Ser397) Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput ELISA platforms that require custom or specialized antibody labeling, such as fluorophores, lanthanides, beads, or biotin. This kit contains capture and detection antibodies that bind to non-overlapping epitopes and is compatible with platforms including MSD, Quanterix Simoa, Alpha Technology (AlphaScreen, AlphaLISA, LANCE, HTRF), and Luminex.
  • Includes 100 μg each of Phospho-YAP (Ser397) (D1E7Y) rabbit monoclonal antibody and YAP (1A12) mouse monoclonal antibody.
  • Formulated in 1X PBS (pH 7.8), free of BSA and azide.
  • Optimized for human species reactivity.
  • Supplied as a liquid in a clear appearance.
  • Recommended for use in ELISA and related immunoassay applications.