Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Phospho-Zap-70 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP

Catalog No. 503131934
Encompass

This matched antibody pair is designed for immunoassay and high-throughput ELISA platforms requiring custom labeling, including fluorophores, lanthanides, biotin, and beads. The kit provides capture and detection antibodies that bind to non-overlapping epitopes, supporting various detection platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. It is supplied in a BSA and azide-free formulation in PBS.

  • Ideal for high-throughput immunoassay technologies
  • Compatible with diverse labeling methods and detection platforms
  • Contains capture and detection antibodies targeting ZAP70 (Tyr319) phosphate
  • Formulated in 1X PBS (pH 7.8)
  • BSA and azide free
  • Species reactivity is limited to human

Catalog No. 50-313-1934 Supplier Cell Signaling Technology Supplier No. 26501S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This matched antibody pair is designed for immunoassay and high-throughput ELISA platforms requiring custom labeling, including fluorophores, lanthanides, biotin, and beads. The kit provides capture and detection antibodies that bind to non-overlapping epitopes, supporting various detection platforms such as MSD, Quanterix Simoa, Alpha Technology, and Luminex. It is supplied in a BSA and azide-free formulation in PBS.

  • Ideal for high-throughput immunoassay technologies
  • Compatible with diverse labeling methods and detection platforms
  • Contains capture and detection antibodies targeting ZAP70 (Tyr319) phosphate
  • Formulated in 1X PBS (pH 7.8)
  • BSA and azide free
  • Species reactivity is limited to human

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.