Learn More
Aviva Systems Biology Corp PHOSPHO2 Antibody - N-terminal region (ARP40264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHOSPHO2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PHOSPHO2. Alternate names: MGC111048, MGC22679ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-284-861