Learn More
Cell Biolabs Inc PhosphoBLOCKER™ Blocking Reagent, 200 g dry blend (4 Liters prepared)
Catalog No. NC0247892
Encompass_Preferred
The utilization of anti-phosphoprotein antibodies in western blotting has become a commonly used tool for signal transduction research. Unfortunately, low levels of endogenous phosphoprotein in various cell lysates often can not be detected, even with high concentrations of antibody and long exposure times. Most commercially available western blot blockers (e.g. dry milk, serum) are sufficient to block the unreacted sites on the membrane, reducing the amount of nonspecific antibody binding during the assay; however, they are not designed to preserve phosphoprotein antigens during blotting. Cell Biolabs’ PhosphoBLOCKER™ contains a proprietary formulation that provides several advantages over conventional blockers: (1) It is designed specifically for phosphoprotein blotting; (2) It enhances low level phosphoprotein signal without increasing background; and (3) it is an easy to use premixed dry blend. 200 grams (reconstituted to 4 liters).
Catalog No. NC0247892 Supplier Cell Biolabs Inc Supplier No. AKR104
The utilization of anti-phosphoprotein antibodies in western blotting has become a commonly used tool for signal transduction research. Unfortunately, low levels of endogenous phosphoprotein in various cell lysates often can not be detected, even with high concentrations of antibody and long exposure times. Most commercially available western blot blockers (e.g. dry milk, serum) are sufficient to block the unreacted sites on the membrane, reducing the amount of nonspecific antibody binding during the assay; however, they are not designed to preserve phosphoprotein antigens during blotting. Cell Biolabs’ PhosphoBLOCKER™ contains a proprietary formulation that provides several advantages over conventional blockers: (1) It is designed specifically for phosphoprotein blotting; (2) It enhances low level phosphoprotein signal without increasing background; and (3) it is an easy to use premixed dry blend. 200 grams (reconstituted to 4 liters).