Learn More
Medchemexpress LLC Phosphocreatine (disodium hydrate) | 19333-65-4 | 99.6% | 1 G
SDP
Catalog No. 5000409876
Encompass_Preferred
Phosphocreatine (disodium hydrate) is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart. It normalizes mitochondrial function and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. This compound also provides renal protection by suppressing apoptosis and reactive oxygen species generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Protects against doxorubicin-induced cardiomyocyte injury by targeting TAK1.
  • Alleviates oxidative stress by increasing antioxidant activity.
  • Attenuates cell injury and inhibits apoptosis induced by methylglyoxal.
  • Prevents loss of mitochondrial membrane permeability.
  • Exhibits neuroprotective effects by normalizing mitochondrial function.
  • Protects kidney tissues against diabetic nephropathy.
Catalog No. 50-004-09876 Supplier Medchemexpress LLC Supplier No. HYD0885D1G
Phosphocreatine (disodium hydrate) is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart. It normalizes mitochondrial function and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. This compound also provides renal protection by suppressing apoptosis and reactive oxygen species generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Protects against doxorubicin-induced cardiomyocyte injury by targeting TAK1.
  • Alleviates oxidative stress by increasing antioxidant activity.
  • Attenuates cell injury and inhibits apoptosis induced by methylglyoxal.
  • Prevents loss of mitochondrial membrane permeability.
  • Exhibits neuroprotective effects by normalizing mitochondrial function.
  • Protects kidney tissues against diabetic nephropathy.