Learn More
Medchemexpress LLC Phosphocreatine disodium hydrate | 19333-65-4 | 99.6% | 500 MG
SDP
Catalog No. 5000410690
Encompass_Preferred
Phosphocreatine disodium hydrate is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart, normalizes mitochondrial function, and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. It also provides renal protection by suppressing apoptosis and reactive oxygen species generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Enhances antioxidant activity
  • Activates TAK1 pathway to protect the heart
  • Normalizes mitochondrial function
  • Reduces oxidative stress
  • Provides renal protection
Catalog No. 50-004-10690 Supplier Medchemexpress LLC Supplier No. HYD0885D500MG
Phosphocreatine disodium hydrate is an organic compound found in vertebrate skeletal muscles. It enhances antioxidant activity and activates the TAK1 pathway to protect the heart, normalizes mitochondrial function, and reduces oxidative stress via the Akt-mediated Nrf2/HO-1 pathway. It also provides renal protection by suppressing apoptosis and reactive oxygen species generation through the ERK-mediated Nrf-2/HO-1 pathway.
  • Enhances antioxidant activity
  • Activates TAK1 pathway to protect the heart
  • Normalizes mitochondrial function
  • Reduces oxidative stress
  • Provides renal protection