Learn More
Worthington Biochemical Corporation PHOSPHODIESTERASE II
SDP
Catalog No. NC0428520
Encompass
Prepared from the 1mM sodium pyrophosphate, pH 6.9, alumina gel eluate of Hilmoe: Biochem. Prep VIII (Meister, A., ed.), John Wiley & Sons, NY, NY, p. 105 (1961). Lyophilized in vials.
Catalog No. NC0428520 Supplier Worthington Biochemical Corporation Supplier No. LS003602
Prepared from the 1mM sodium pyrophosphate, pH 6.9, alumina gel eluate of Hilmoe: Biochem. Prep VIII (Meister, A., ed.), John Wiley & Sons, NY, NY, p. 105 (1961). Lyophilized in vials.