Learn More
Medchemexpress LLC Phosphodiesterase II, Bovine Spleen (3′-Exonuclease) | 9068-54-6 | 10 U
SDP
Catalog No. 5000397462
Encompass_Preferred
Phosphodiesterase II (EC 3.1.16.1), also referred to as phosphodiesterase 2, plays a primary role in the hydrolysis of crucial second messengers, cyclic adenosine monophosphate (cAMP) and cyclic guanosine monophosphate (cGMP), and is frequently utilized in biochemical research. This enzyme is expressed in a variety of tissues, such as the adrenal medulla, brain, heart, platelets, macrophages, and endothelial cells, and is implicated in the regulation of many different intracellular processes.
  • Appearance: solid
  • Color: white to off-white
  • Enzyme activity: ≥1.20 U/mg solid
  • For research use only
Catalog No. 50-003-97462 Supplier Medchemexpress LLC Supplier No. HYP286710U
Phosphodiesterase II (EC 3.1.16.1), also referred to as phosphodiesterase 2, plays a primary role in the hydrolysis of crucial second messengers, cyclic adenosine monophosphate (cAMP) and cyclic guanosine monophosphate (cGMP), and is frequently utilized in biochemical research. This enzyme is expressed in a variety of tissues, such as the adrenal medulla, brain, heart, platelets, macrophages, and endothelial cells, and is implicated in the regulation of many different intracellular processes.
  • Appearance: solid
  • Color: white to off-white
  • Enzyme activity: ≥1.20 U/mg solid
  • For research use only